Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006492.2 | NP_006483.2 | 2 | 615 | ||
GenBank mRNA | AF008203.1 | - | 2 | 527 | ||
BC112007.1 | - | 2 | 615 | 8327475 | ||
BC113428.1 | - | 2 | 615 | 8322480 |