Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005073.3 | NP_005064.1 | 20 | 1736 | ||
GenBank mRNA | AF043233.1 | - | 20 | 1715 | ||
AY634368.1 | - | 20 | 1706 | |||
BC096327.1 | - | 20 | 1690 | 40008278 | ||
BC096328.3 | - | 20 | 1690 | 40008279 | ||
BC096329.3 | - | 20 | 1690 | 40008280 | ||
EU544224.1 | - | 19 | 1659 | |||
U13173.1 | - | 20 | 1715 | |||
U21936.1 | - | 20 | 1715 |