Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013191.1 | NP_037323.1 | 2 | 187 | ||
XM_008772868.1 | XP_008771090.1 | 2 | 186 | |||
GenBank mRNA | BC087026.1 | - | 2 | 125 | 7193740 | |
FQ213687.1 | - | 2 | 179 | |||
FQ213774.1 | - | 2 | 181 | |||
FQ213840.1 | - | 2 | 137 | |||
FQ214218.1 | - | 2 | 131 | |||
FQ214292.1 | - | 2 | 160 | |||
FQ214554.1 | - | 2 | 129 | |||
M15705.1 | - | 1 | 54 | |||
M54919.1 | - | 2 | 187 | |||
X01090.1 | - | 2 | 187 |