Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013197.1 | NP_037329.1 | 8 | 1269 | ||
GenBank mRNA | BC081857.1 | - | 1290 | 7133382 | ||
D86297.1 | - | 8 | 1269 |