Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rheb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013216.1 | NP_037348.1 | 4 | 345 | ||
GenBank mRNA | BC088857.1 | - | 4 | 399 | 7307969 | |
FQ212847.1 | - | 4 | 367 | |||
FQ214530.1 | - | 4 | 384 | |||
FQ218725.1 | - | 4 | 367 | |||
FQ225701.1 | - | 4 | 349 | |||
FQ230203.1 | - | 4 | 349 | |||
U08227.1 | - | 4 | 345 |