Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013221.2 | NP_037353.2 | 8, 9 | 1115 | ||
XM_006239975.2 | XP_006240037.1 | 8, 9 | 1178 | |||
GenBank mRNA | BC091165.1 | - | 7, 8 | 1089 | 7097013 | |
FQ223257.1 | - | 8, 9 | 1222 | |||
FQ224288.1 | - | 8, 9 | 1106 | |||
U09551.1 | - | 7, 8 | 1110 |