Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005074.3 | NP_005065.2 | 9 | 1107 | ||
XM_011514818.2 | XP_011513120.2 | 9 | 1144 | |||
XM_011514819.2 | XP_011513121.2 | 8 | 1057 | |||
XM_011514820.2 | XP_011513122.2 | 8 | 982 | |||
XM_011514821.2 | XP_011513123.1 | 7 | 861 | |||
XM_017011199.1 | XP_016866688.1 | 9 | 1144 | |||
XM_017011200.1 | XP_016866689.1 | 9 | 1144 | |||
XM_017011201.1 | XP_016866690.1 | 9 | 1144 | |||
XM_017011202.1 | XP_016866691.1 | 8 | 1072 | |||
GenBank mRNA | AK290783.1 | - | 9 | 1089 | ||
AK308035.1 | - | 9 | 1089 | |||
AY780791.1 | - | 7 | 829 | |||
BC101745.1 | - | 8 | 1044 | 8069251 | ||
BC101747.1 | - | 8 | 1044 | 8069253 | ||
D28532.1 | - | 8 | 988 | |||
X71355.1 | - | 8 | 1003 |