Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC34A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003052.4 | NP_003043.3 | 13 | 1552 | ||
XM_005265975.1 | XP_005266032.1 | 11 | 1327 | |||
XM_017009774.1 | XP_016865263.1 | 6 | 650 | |||
GenBank mRNA | AK096088.1 | - | 13 | 1531 | ||
BC050385.1 | - | 13 | 1529 | 5760855 | ||
BC053349.1 | - | 13 | 1539 | 5186103 | ||
L13258.1 | - | 13 | 1529 |