Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191810.1 | NP_001178739.1 | 18 | 1526 | ||
XM_008767220.1 | XP_008765442.1 | 18 | 1566 |