Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019149.3 | NP_062022.2 | 2 | 876 | ||
XM_006254534.2 | XP_006254596.1 | 2 | 898 | |||
XM_006254535.2 | XP_006254597.1 | 2 | 849 | |||
GenBank mRNA | AB205483.1 | - | 1 | 1646 | ||
BC062231.1 | - | 2 | 899 | |||
BC168723.1 | - | 2 | 848 | |||
M63485.1 | - | 2 | 861 |