Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC18A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003054.4 | NP_003045.2 | 3 | 304 | ||
GenBank mRNA | AK314997.1 | - | 3 | 257 | ||
BC030593.1 | - | 3 | 289 | |||
BC108927.1 | - | 2 | 169 | 40025174 | ||
BC108928.2 | - | 2 | 169 | 40025175 | ||
DB458870.1 | - | 3 | 304 | |||
L09118.1 | - | 3 | 246 | |||
L14269.1 | - | 3 | 174 | |||
L23205.1 | - | 3 | 254 | |||
X71354.1 | - | 3 | 204 |