Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031560.2 | NP_113748.1 | 8 | 1011 | ||
GenBank mRNA | AF010306.1 | - | 8 | 980 | ||
BC078793.1 | - | 8 | 1011 | 7134253 | ||
FQ222438.1 | - | 8 | 955 | |||
FQ228691.1 | - | 8 | 1028 |