Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168631.1 | NP_001162102.1 | 7 | 1437 | ||
XM_006232079.2 | XP_006232141.1 | 7 | 1513 | |||
XM_006232080.2 | XP_006232142.1 | 7 | 1545 |