Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inhba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017128.2 | NP_058824.1 | 3 | 693 | ||
XM_006254001.2 | XP_006254063.1 | 4 | 4352 | |||
XM_008771712.1 | XP_008769934.1 | 3 | 4270 | |||
XM_008771713.1 | XP_008769935.1 | 5 | 6060 | |||
XM_008771714.1 | XP_008769936.1 | 5 | 2840 | |||
XM_008771715.1 | XP_008769937.1 | 5 | 2742 | |||
XM_008771716.1 | XP_008769938.1 | 5 | 1120 | |||
GenBank mRNA | M37482.1 | - | 3 | 685 |