Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256534.1 | NP_001243463.1 | 7 | 933 | ||
NM_001287387.1 | NP_001274316.1 | 8 | 787 | |||
NM_005984.4 | NP_005975.1 | 8 | 952 | |||
NR_046298.2 | 7 | 813 | ||||
GenBank mRNA | AK292313.1 | - | 8 | 856 | ||
AK298204.1 | - | 8 | 785 | |||
BC004980.1 | - | 8 | 813 | 2957917 | ||
BC008061.2 | - | 8 | 815 | 2958945 | ||
BC018590.1 | - | 8 | 710 | |||
BI160240.1 | - | 7 | 677 | |||
HM037273.1 | - | 7 | 788 | |||
L75823.1 | - | 8 | 861 | |||
L77567.1 | - | 8 | 834 | |||
U25147.1 | - | 8 | 836 |