Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021740.1 | NP_068508.1 | 5 | 1080 | ||
GenBank mRNA | BC061972.1 | - | 5 | 1092 | 5623463 | |
BC092569.1 | - | 5 | 1104 | 7307282 | ||
FQ210626.1 | - | 5 | 1113 | |||
FQ214221.1 | - | 5 | 1108 | |||
FQ215633.1 | - | 5 | 1113 | |||
FQ219686.1 | - | 5 | 1114 | |||
FQ220677.1 | - | 5 | 1113 | |||
FQ221801.1 | - | 5 | 1113 | |||
FQ225103.1 | - | 5 | 1117 | |||
FQ226946.1 | - | 5 | 1114 | |||
FQ227835.1 | - | 5 | 1115 | |||
FQ229930.1 | - | 5 | 1113 | |||
FQ231794.1 | - | 5 | 1119 | |||
FQ232039.1 | - | 5 | 1119 | |||
M86564.1 | - | 5 | 1080 | |||
X55326.1 | - | 5 | 1080 |