Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005630.2 | NP_005621.2 | 9, 10 | 1577 | ||
XM_017007075.1 | XP_016862564.1 | 9, 10 | 1520 | |||
XM_017007076.1 | XP_016862565.1 | 9, 10 | 1520 | |||
XM_017007077.1 | XP_016862566.1 | 10, 11 | 1565 | |||
GenBank mRNA | AK303550.1 | - | 5, 6 | 883 | ||
BC041140.1 | - | 9, 10 | 1406 | 5751516 | ||
BC051347.1 | - | 9, 10 | 1486 | 6481104 | ||
U70867.1 | - | 9, 10 | 1386 |