Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021094.3 | NP_066580.1 | 9 | 1423 | ||
NM_134431.3 | NP_602307.1 | 11 | 1893 | |||
XM_005253474.3 | XP_005253531.1 | 10 | 1314 | |||
XM_005253477.3 | XP_005253534.1 | 9 | 1201 | |||
XM_011520818.1 | XP_011519120.1 | 10 | 1321 | |||
XM_011520819.1 | XP_011519121.1 | 10 | 1468 | |||
XM_011520820.2 | XP_011519122.1 | 10 | 2790 | |||
XM_011520821.2 | XP_011519123.1 | 3 | 1593 | |||
XM_017019849.1 | XP_016875338.1 | 10 | 1321 | |||
XM_017019850.1 | XP_016875339.1 | 9 | 1201 | |||
GenBank mRNA | AF085224.1 | - | 10 | 1382 | ||
AF279784.1 | - | 11 | 1893 | |||
AK296044.1 | - | 8 | 1034 | |||
AK316249.1 | - | 9 | 1453 | |||
BC042452.1 | - | 9 | 1203 | 4820050 | ||
U21943.1 | - | 9 | 1225 |