Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019174.3 | NP_062047.1 | 4 | 499 | ||
GenBank mRNA | BC097329.1 | - | 4 | 394 | 7445156 | |
CB746957.1 | - | 1 | 60 | |||
FM062617.1 | - | 4 | 499 | |||
FQ222059.1 | - | 449 | ||||
S68245.1 | - | 4 | 442 |