Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021094.3 | NP_066580.1 | 8 | 1270 | ||
NM_134431.3 | NP_602307.1 | 10 | 1740 | |||
XM_005253474.3 | XP_005253531.1 | 9 | 1161 | |||
XM_005253477.3 | XP_005253534.1 | 8 | 1048 | |||
XM_011520818.1 | XP_011519120.1 | 9 | 1168 | |||
XM_011520819.1 | XP_011519121.1 | 9 | 1315 | |||
XM_011520820.2 | XP_011519122.1 | 9 | 2637 | |||
XM_011520821.2 | XP_011519123.1 | 2 | 1440 | |||
XM_017019849.1 | XP_016875338.1 | 9 | 1168 | |||
XM_017019850.1 | XP_016875339.1 | 8 | 1048 | |||
GenBank mRNA | AF085224.1 | - | 9 | 1229 | ||
AF279784.1 | - | 10 | 1740 | |||
AK296044.1 | - | 7 | 881 | |||
AK316249.1 | - | 8 | 1300 | |||
BC042452.1 | - | 8 | 1050 | 4820050 | ||
U21943.1 | - | 8 | 1072 |