Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057183.1 | NP_476531.1 | 3 | 321 | ||
GenBank mRNA | DV216119.1 | - | 3 | 321 | ||
FQ233430.1 | - | 3 | 348 | |||
L05175.1 | - | 3 | 264 |