Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031569.3 | NP_113757.1 | 2 | 318 | ||
XM_006235736.2 | XP_006235798.1 | 3 | 408 | |||
XM_006235738.2 | XP_006235800.1 | 2 | 166 | |||
XM_006235739.2 | XP_006235801.1 | 3 | 408 | |||
GenBank mRNA | AF115267.1 | - | 1 | 105 | ||
AY152731.1 | - | 1 | 77 | |||
D16438.1 | - | 2 | 129 | |||
L28144.1 | - | 2 | 249 | |||
L29419.1 | - | 1 | 212 | |||
L33916.1 | - | 2 | 183 | |||
U01913.1 | - | 2 | 258 | |||
U05239.1 | - | 2 | 260 | |||
U07871.1 | - | 1 | 108 |
Set Membership: |
G-protein coupled receptor |