Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019177.3 | NP_062050.3 | 6 | 1040 | ||
GenBank mRNA | D10831.1 | - | 6 | 1032 | ||
S79523.1 | - | 6 | 1040 |