Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017145.2 | NP_058841.1 | 4 | 498 | ||
GenBank mRNA | AF063851.1 | - | 4 | 499 | ||
FQ220114.1 | - | 4 | 498 | |||
FQ220424.1 | - | 4 | 460 | |||
FQ220659.1 | - | 4 | 498 | |||
FQ220808.1 | - | 4 | 531 | |||
FQ221313.1 | - | 4 | 498 | |||
FQ228378.1 | - | 4 | 498 | |||
FQ229265.1 | - | 4 | 498 | |||
FQ229320.1 | - | 4 | 498 | |||
FQ229582.1 | - | 4 | 495 | |||
FQ229760.1 | - | 4 | 498 |