Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019181.1 | NP_062054.1 | 2 | 469 | ||
GenBank mRNA | BC166809.1 | - | 2 | 469 | 8373837 | |
DY309412.1 | - | 2 | 467 | |||
DY314075.1 | - | 2 | 468 | |||
FQ226380.1 | - | 2 | 306 | |||
FQ227075.1 | - | 2 | 319 | |||
M36410.1 | - | 2 | 445 |
Gene Symbol: RGD1560795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236796.2 | XP_006236858.1 | 1 | 257 |