Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019183.1 | NP_062056.1 | 4, 5 | 814 | ||
GenBank mRNA | BC127451.1 | - | 4, 5 | 689 | 7379877 | |
CF109672.1 | - | 4, 5 | 668 | |||
X80130.1 | - | 4, 5 | 700 |