Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019184.2 | NP_062057.2 | 7 | 1000 | ||
XM_008760364.1 | XP_008758586.1 | 6 | 828 | |||
GenBank mRNA | BC088146.1 | - | 7 | 990 | 7369647 | |
J02657.1 | - | 7 | 1000 | |||
U33173.1 | - | 7 | 1000 |