Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007599.2 | NP_001007600.1 | 4 | 279 | ||
GenBank mRNA | BC058500.1 | - | 4 | 283 | 6921061 | |
FQ216928.1 | - | 4 | 285 | |||
FQ221252.1 | - | 4 | 285 | |||
FQ221690.1 | - | 4 | 309 | |||
FQ221693.1 | - | 4 | 285 | |||
FQ222524.1 | - | 4 | 285 | |||
FQ222678.1 | - | 4 | 285 | |||
FQ223359.1 | - | 4 | 285 | |||
FQ224963.1 | - | 4 | 285 | |||
FQ228675.1 | - | 4 | 286 | |||
X58200.1 | - | 4 | 277 |