Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001633.3 | NP_001624.1 | 1, 2 | 376 | ||
GenBank mRNA | AK290837.1 | - | 1, 2 | 204 | ||
AK310776.1 | - | 1, 2 | 303 | |||
AK310791.1 | - | 1, 2 | 232 | |||
AY544123.1 | - | 1, 2 | 173 | |||
BC041593.1 | - | 1, 2 | 339 | 5433460 | ||
CB141734.1 | - | 1, 2 | 376 | |||
X04225.1 | - | 1, 2 | 185 | |||
X04494.1 | - | 1, 2 | 158 |