Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053302.3 | NP_445754.2 | 2 | 767 | ||
XM_008765379.1 | XP_008763601.1 | 2 | 1081 | |||
XM_008765380.1 | XP_008763602.1 | 1 | 1131 | |||
GenBank mRNA | BC129072.1 | - | 2 | 767 | 7096756 | |
S79811.1 | - | 2 | 845 |
Set Membership: |
G-protein coupled receptor |