Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012459.1 | NP_001012477.1 | 2 | 317 | ||
GenBank mRNA | AB009246.1 | - | 2 | 199 | ||
BC090344.1 | - | 2 | 317 | 7304238 |