Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003064.3 | NP_003055.1 | 3 | 289 | ||
GenBank mRNA | AF114471.1 | - | 3 | 268 | ||
AK312192.1 | - | 3 | 273 | |||
AX772818.1 | - | 3 | 289 | |||
BC020708.1 | - | 3 | 289 | 4733996 | ||
X04470.1 | - | 3 | 286 | |||
X04503.1 | - | 3 | 268 |