Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uncx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017179.1 | NP_058875.1 | 3 | 1120 | ||
GenBank mRNA | D87748.1 | - | 3 | 1120 |