Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001633.3 | NP_001624.1 | 8 | 1077 | ||
GenBank mRNA | AK290837.1 | - | 8 | 905 | ||
AK303795.1 | - | 8 | 803 | |||
AY544123.1 | - | 8 | 874 | |||
BC041593.1 | - | 8 | 1040 | 5433460 | ||
X04225.1 | - | 8 | 886 | |||
X04494.1 | - | 8 | 859 |