Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017186.2 | NP_058882.1 | 3 | 437 | ||
GenBank mRNA | AF081557.1 | - | 3 | 438 |