Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289396.1 | NP_001276325.1 | 2, 3 | 442 | ||
NM_001289397.1 | NP_001276326.1 | 2, 3 | 455 | |||
NM_003070.4 | NP_003061.3 | 2, 3 | 455 | |||
NM_139045.3 | NP_620614.2 | 2, 3 | 455 | |||
GenBank mRNA | AK296373.1 | - | 2, 3 | 396 | ||
AK299683.1 | - | 2, 3 | 362 | |||
BC040029.1 | - | 2, 3 | 332 | |||
BM921013.1 | - | 396 | ||||
X72889.1 | - | 2, 3 | 455 |