Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017198.1 | NP_058894.1 | 13 | 1618 | ||
XM_006229752.1 | XP_006229814.1 | 13 | 1469 | |||
XM_006229753.1 | XP_006229815.1 | 13 | 1466 | |||
XM_006229754.2 | XP_006229816.1 | 13 | 1458 | |||
GenBank mRNA | U23443.1 | - | 13 | 1424 | ||
U49953.1 | - | 13 | 1618 |