Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Por | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031576.1 | NP_113764.1 | 103 | |||
GenBank mRNA | M10068.1 | - | 1 | 67 | ||
M12516.1 | - | 103 |