Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017203.1 | NP_058899.1 | 3 | 1180 | ||
GenBank mRNA | AF092733.1 | - | 3 | 985 |