Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019217.1 | NP_062090.1 | 5 | 6717 | ||
XM_008760661.1 | XP_008758883.1 | 4 | 6382 | |||
GenBank mRNA | CB583564.1 | - | 1 | 312 | ||
X16623.1 | - | 1 | 1932 | |||
X60370.1 | - | 3 | 6262 |