Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019219.2 | NP_062092.2 | 4, 5 | 350 | ||
GenBank mRNA | AF025819.2 | - | 4, 5 | 350 | ||
BC101915.1 | - | 4, 5 | 394 | 7935765 |