Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019220.1 | NP_062093.1 | 3 | 304 | ||
GenBank mRNA | BC101855.1 | - | 269 | 7933368 | ||
L14462.1 | - | 3 | 304 |