Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMD1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC018676.1 | - | 1 | 195 |
Gene Symbol: AMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287214.1 | NP_001274143.1 | 5 | 800 | ||
NM_001287215.1 | NP_001274144.1 | 6 | 650 | |||
NM_001287216.1 | NP_001274145.1 | 2 | 527 | |||
NM_001634.5 | NP_001625.2 | 6 | 887 | |||
NR_109768.1 | 3 | 570 | ||||
GenBank mRNA | AJ420527.1 | - | 3 | 545 | ||
AK125644.1 | - | 2 | 1510 | |||
AK300330.1 | - | 5 | 799 | |||
AK302761.1 | - | 2 | 528 | |||
AK304351.1 | - | 6 | 650 | |||
AK316471.1 | - | 6 | 519 | |||
AL832698.1 | - | 3 | 569 | |||
BC000171.2 | - | 6 | 660 | 2900587 | ||
BX640599.1 | - | 6 | 887 | |||
M21154.1 | - | 6 | 813 |