Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003076.4 | NP_003067.3 | 5, 6 | 830 | ||
NM_139071.2 | NP_620710.2 | 5, 6 | 830 | |||
XM_011538695.1 | XP_011536997.1 | 5, 6 | 830 | |||
GenBank mRNA | AF109733.1 | - | 5, 6 | 618 | ||
AK130517.1 | - | 6, 7 | 569 | |||
AK293932.1 | - | 5, 6 | 688 | |||
BC009368.2 | - | 5, 6 | 780 | 4110499 | ||
CN272379.1 | - | 4, 5 | 360 | |||
DA825057.1 | - | 1, 2 | 101 | |||
U66617.1 | - | 5, 6 | 808 |