Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017212.2 | NP_058908.2 | 6, 7 | 769 | ||
XM_008768268.1 | XP_008766490.1 | 10, 11 | 1942 | |||
XM_008768269.1 | XP_008766491.1 | 9, 10 | 1855 | |||
XM_008768271.1 | XP_008766493.1 | 8, 9 | 1768 | |||
XM_008768272.1 | XP_008766494.1 | 9, 10 | 1743 | |||
XM_008768274.1 | XP_008766496.1 | 9, 10 | 1176 | |||
XM_008768275.1 | XP_008766497.1 | 8, 9 | 1089 | |||
XM_008768277.1 | XP_008766499.1 | 8, 9 | 975 | |||
XM_008768278.1 | XP_008766500.1 | 7, 8 | 888 | |||
XM_008768282.1 | XP_008766504.1 | 11, 12 | 1997 | |||
GenBank mRNA | CB615367.1 | - | 3, 4 | 432 | ||
M84156.1 | - | 9, 10 | 1600 | |||
X79321.1 | - | 6, 7 | 856 |