Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017212.2 | NP_058908.2 | 6 | 516 | ||
XM_008768268.1 | XP_008766490.1 | 10 | 1689 | |||
XM_008768269.1 | XP_008766491.1 | 9 | 1602 | |||
XM_008768270.1 | XP_008766492.1 | 10 | 1690 | |||
XM_008768271.1 | XP_008766493.1 | 8 | 1515 | |||
XM_008768272.1 | XP_008766494.1 | 9 | 1490 | |||
XM_008768273.1 | XP_008766495.1 | 9 | 1490 | |||
XM_008768274.1 | XP_008766496.1 | 9 | 923 | |||
XM_008768275.1 | XP_008766497.1 | 8 | 836 | |||
XM_008768277.1 | XP_008766499.1 | 8 | 722 | |||
XM_008768278.1 | XP_008766500.1 | 7 | 635 | |||
XM_008768279.1 | XP_008766501.1 | 8 | 723 | |||
XM_008768280.1 | XP_008766502.1 | 7 | 635 | |||
XM_008768281.1 | XP_008766503.1 | 6 | 548 | |||
XM_008768282.1 | XP_008766504.1 | 11 | 1744 | |||
GenBank mRNA | BC126095.1 | - | 6 | 516 | 8367931 | |
CB581821.1 | - | 6 | 496 | |||
CB615367.1 | - | 3 | 179 | |||
DY311887.1 | - | 6 | 516 | |||
M84156.1 | - | 9 | 1347 | |||
X79321.1 | - | 6 | 603 |