Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003076.4 | NP_003067.3 | 9 | 1281 | ||
NM_139071.2 | NP_620710.2 | 9 | 1281 | |||
XM_005269107.3 | XP_005269164.2 | 8 | 1158 | |||
GenBank mRNA | AF109733.1 | - | 9 | 1069 | ||
AK130517.1 | - | 10 | 1021 | |||
AK292700.1 | - | 9 | 945 | |||
BC009368.2 | - | 9 | 1231 | 4110499 | ||
DA825057.1 | - | 5 | 553 | |||
U66617.1 | - | 9 | 1259 |