Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019224.2 | NP_062097.1 | 15 | 1237 | ||
GenBank mRNA | AB019145.1 | - | 15 | 1237 | ||
AF038006.1 | - | 6 | 563 | |||
AF071475.1 | - | 15 | 1233 | |||
U32433.1 | - | 2 | 127 |