Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003076.4 | NP_003067.3 | 9, 10 | 1311 | ||
NM_139071.2 | NP_620710.2 | 9, 10 | 1311 | |||
XM_005269107.3 | XP_005269164.2 | 8, 9 | 1188 | |||
GenBank mRNA | AF109733.1 | - | 9, 10 | 1099 | ||
AK130517.1 | - | 10, 11 | 1051 | |||
AK292700.1 | - | 9, 10 | 975 | |||
BC009368.2 | - | 9, 10 | 1261 | 4110499 | ||
N75126.1 | - | 21 | ||||
U66617.1 | - | 9, 10 | 1289 |