Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289941.1 | NP_001276870.1 | 6 | 1048 | ||
NM_001289942.1 | NP_001276871.1 | 7 | 1175 | |||
NM_001289943.1 | NP_001276872.1 | 5 | 839 | |||
NM_019225.2 | NP_062098.1 | 7 | 1165 | |||
GenBank mRNA | AF265360.1 | - | 6 | 887 | ||
S59158.1 | - | 7 | 1107 |